ENA508624774 Enamine

4-(morpholin-4-yl)oxane-4-carbaldehyde

  • MDL number MFCD29991316
  • Cas Number : 1888919-78-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-260185

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-260185
Availability details for
Quantity
  • Availability
  • Add to Cart