ENA508624785 Enamine

4-(3-bromo-2-oxopropyl)benzonitrile

  • MDL number MFCD28808405
  • Cas Number : 1196532-44-1
  • Molecular Weight : 238.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-260807

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-260807
Availability details for
Quantity
  • Availability
  • Add to Cart