ENA508624821 Enamine

methyl 2-methyl-1-(propan-2-yl)-4,5-dihydro-1H-pyrrole-3-carboxylate

  • MDL number MFCD29991332
  • Cas Number : 116195-42-7
  • Molecular Weight : 183.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-261331

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-261331
Availability details for
Quantity
  • Availability
  • Add to Cart