ENA508624823 Enamine

(2-bromothiophen-3-yl)methanesulfonyl chloride

  • MDL number MFCD29991334
  • Cas Number : 1935091-96-5
  • Molecular Weight : 275.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-261407

Safety Information

signalword Danger
pcodes P260,P264,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P363,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-261407
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart