ENA508624836 Enamine

3-(2-oxopyrrolidin-1-yl)butanoic acid

  • MDL number MFCD11649544
  • Cas Number : 100911-30-6
  • Molecular Weight : 171.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-261653

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-261653
Availability details for
Quantity
  • Availability
  • Add to Cart