ENA508624843 Enamine

1,4-dibromo-2,5-diethoxybenzene

  • MDL number MFCD08696083
  • Cas Number : 101907-55-5
  • Molecular Weight : 324.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-261776

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-261776
Availability details for
Quantity
  • Availability
  • Add to Cart