ENA508624847 Enamine

1-(propan-2-yl)-2-azaspiro[3.4]oct-6-ene hydrochloride

  • MDL number MFCD29991338
  • Cas Number : 1955524-17-0
  • Molecular Weight : 187.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-261862

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-261862
Availability details for
Quantity
  • Availability
  • Add to Cart