ENA508624875 Enamine

2,6-dimethyl-1-(propan-2-yl)piperazine

  • MDL number MFCD09894132
  • Cas Number : 1017366-56-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-262519

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-262519
Availability details for
Quantity
  • Availability
  • Add to Cart