ENA508624877 Enamine

4-acetylpiperazine-1-carbothioamide

  • MDL number MFCD04116668
  • Cas Number : 388118-53-4
  • Molecular Weight : 187.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-262629

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-262629
Availability details for
Quantity
  • Availability
  • Add to Cart