ENA508624894 Enamine

8-({[(tert-butoxy)carbonyl]amino}methyl)-5,6,7,8-tetrahydronaphthalene-2-carboxylic acid

  • MDL number MFCD24537388
  • Cas Number : 165949-89-3
  • Molecular Weight : 305.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-262817

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-262817
Availability details for
Quantity
  • Availability
  • Add to Cart