ENA508624897 Enamine

5-methyl-1,3-dihydro-2,1-benzoxaborol-1-ol

  • MDL number MFCD22415409
  • Cas Number : 905710-76-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-262844

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-262844
Availability details for
Quantity
  • Availability
  • Add to Cart