ENA508624937 Enamine

bicyclo[4.1.0]heptane-2-carboxylic acid

  • MDL number MFCD24499216
  • Molecular Weight : 140.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-124937

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124937
Availability details for
Quantity
  • Availability
  • Add to Cart