ENA508624960 Enamine

1-[3-(trifluoromethoxy)phenyl]cyclopropane-1-carbonitrile

  • MDL number MFCD11036649
  • Cas Number : 1260653-10-8
  • Molecular Weight : 227.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-263578

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-263578
Availability details for
Quantity
  • Availability
  • Add to Cart