ENA508624996 Enamine

N-methoxy-N-methyloxolane-2-carboxamide

  • MDL number MFCD14707543
  • Cas Number : 907961-91-5
  • Molecular Weight : 159.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-131858

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131858
Availability details for
Quantity
  • Availability
  • Add to Cart