ENA508625003 Enamine

5-amino-1-benzyl-1H-1,2,3-triazole-4-carbonitrile

  • MDL number MFCD02752908
  • Cas Number : 20271-35-6
  • Molecular Weight : 199.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-135376

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135376
Availability details for
Quantity
  • Availability
  • Add to Cart