ENA508625008 Enamine

2-[4-(aminomethyl)oxan-4-yl]acetic acid hydrochloride

  • MDL number MFCD29991400
  • Cas Number : 205827-18-5
  • Molecular Weight : 209.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-263901

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-263901
Availability details for
Quantity
  • Availability
  • Add to Cart