ENA508625011 Enamine

(6-chloro-4-methoxypyridin-2-yl)methanol

  • MDL number MFCD23706787
  • Cas Number : 204378-40-5
  • Molecular Weight : 173.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-263910

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-263910
Availability details for
Quantity
  • Availability
  • Add to Cart