ENA508625054 Enamine

1-(methylsulfanyl)cyclopropane-1-carbonitrile

  • MDL number MFCD21127184
  • Cas Number : 35120-11-7
  • Molecular Weight : 113.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-139098

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-139098
Availability details for
Quantity
  • Availability
  • Add to Cart