ENA508625112 Enamine

2-(thiophene-3-carbonyl)butanenitrile

  • MDL number MFCD12115822
  • Cas Number : 1156898-73-5
  • Molecular Weight : 179.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-264195

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-264195
Availability details for
Quantity
  • Availability
  • Add to Cart