ENA508625543 Enamine

tert-butyl 4-(dimethylamino)pyridine-2-carboxylate

  • MDL number MFCD29991539
  • Cas Number : 1955498-61-9
  • Molecular Weight : 222.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-267115

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-267115
Availability details for
Quantity
  • Availability
  • Add to Cart