ENA508625679 Enamine

1-(3-hydrazinylpropyl)-1H-imidazole trihydrochloride

  • MDL number MFCD29991697
  • Cas Number : 1955547-67-7
  • Molecular Weight : 249.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 3HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-270112

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-270112
Availability details for
Quantity
  • Availability
  • Add to Cart