ENA508625886 Enamine

methyl 4-(diethyl phosphono)-3-methylbut-2-enoate

  • Cas Number : 19945-56-3
  • Molecular Weight : 250.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-269368

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-269368
Availability details for
Quantity
  • Availability
  • Add to Cart