ENA508625896 Enamine

3-benzyl-2,2-dimethylpyrrolidine

  • MDL number MFCD16738137
  • Cas Number : 1249614-40-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-269381

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-269381
Availability details for
Quantity
  • Availability
  • Add to Cart