ENA508625944 Enamine

3-chloro-4-[(3,4-difluorophenyl)methoxy]benzoic acid

  • MDL number MFCD17169528
  • Cas Number : 1355636-71-3
  • Molecular Weight : 298.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-269520

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-269520
Availability details for
Quantity
  • Availability
  • Add to Cart