ENA508625964 Enamine

2,4-dichloro-5,7-difluoro-3-methylquinoline

  • MDL number MFCD20527931
  • Cas Number : 1259439-39-8
  • Molecular Weight : 248.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-269558

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P312,P330,P363,P501
MSDS https://www.enaminestore.com/catalog/EN300-269558
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart