ENA508626051 Enamine

spiro[2.5]octane-5-carboxylic acid

  • MDL number MFCD20230621
  • Cas Number : 1314390-66-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-269809

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-269809
Availability details for
Quantity
  • Availability
  • Add to Cart