ENA508626164 Enamine

ethyl 4-amino-3-[(propan-2-yl)amino]benzoate

  • MDL number MFCD24306309
  • Cas Number : 1557797-41-7
  • Molecular Weight : 222.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-270118

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-270118
Availability details for
Quantity
  • Availability
  • Add to Cart