ENA508626230 Enamine

2-methyl-4-(propan-2-yloxy)piperidine

  • MDL number MFCD29991724
  • Cas Number : 1592650-37-7
  • Molecular Weight : 157.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-270429

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-270429
Availability details for
Quantity
  • Availability
  • Add to Cart