ENA508626245 Enamine

2-(2-fluorophenyl)ethane-1-sulfonamide

  • MDL number MFCD26064749
  • Cas Number : 919354-05-5
  • Molecular Weight : 203.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-283831

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-283831
Availability details for
Quantity
  • Availability
  • Add to Cart