ENA508626268 Enamine

1-[4-(1-phenylethoxy)phenyl]ethan-1-one

  • MDL number MFCD11524181
  • Cas Number : 117321-40-1
  • Molecular Weight : 240.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-295150

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-295150
Availability details for
Quantity
  • Availability
  • Add to Cart