ENA508626462 Enamine

[1,3]oxazolo[5,4-d]pyrimidin-7-amine

  • MDL number MFCD19206076
  • Cas Number : 10325-61-8
  • Molecular Weight : 136.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-297887

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-297887
Availability details for
Quantity
  • Availability
  • Add to Cart