ENA508626757 Enamine

N5-[3-(trifluoromethoxy)phenyl]pyridine-2,5-diamine hydrochloride

  • MDL number MFCD29991857
  • Cas Number : 1955553-42-0
  • Molecular Weight : 305.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-298646

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-298646
Availability details for
Quantity
  • Availability
  • Add to Cart