ENA508626844 Enamine

6-methoxy-1H-indazole-5-carboxamide

  • MDL number MFCD29991959
  • Cas Number : 1955532-15-6
  • Molecular Weight : 191.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-299688

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-299688
Availability details for
Quantity
  • Availability
  • Add to Cart