ENA508626953 Enamine

propan-2-yl N-(4-amino-3-methylbutan-2-yl)carbamate

  • MDL number MFCD29991868
  • Cas Number : 1592253-28-5
  • Molecular Weight : 188.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-298813

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-298813
Availability details for
Quantity
  • Availability
  • Add to Cart