ENA508626975 Enamine

benzyl N-[2-(difluoromethoxy)ethyl]carbamate

  • MDL number MFCD29991877
  • Cas Number : 1919864-85-9
  • Molecular Weight : 245.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-298850

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-298850
Availability details for
Quantity
  • Availability
  • Add to Cart