ENA508627061 Enamine

1-[(3-methoxyphenyl)methyl]piperidin-4-amine dihydrochloride

  • MDL number MFCD15200716
  • Cas Number : 57645-53-1
  • Molecular Weight : 293.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-299234

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-299234
Availability details for
Quantity
  • Availability
  • Add to Cart