ENA508627120 Enamine

6-phenylimidazo[2,1-b][1,3,4]thiadiazole

  • MDL number MFCD00193132
  • Cas Number : 25752-18-5
  • Molecular Weight : 201.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-298400

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-298400
Availability details for
Quantity
  • Availability
  • Add to Cart