ENA508627297 Enamine

2-ethyl-6-methylpyrazine

  • MDL number MFCD00055035
  • Cas Number : 13925-03-6
  • Molecular Weight : 122.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-300011

Safety Information

signalword Warning
pcodes P210,P233,P240,P241,P242,P243,P264,P270,P280,P301+P312,P303+P361+P353,P330,P370+P378,P403+P235,P501
MSDS https://www.enaminestore.com/catalog/EN300-300011
pictograms GHS02,GHS07
hstatements H226,H302
Availability details for
Quantity
  • Availability
  • Add to Cart