ENA508627452 Enamine

1-(2-{[(tert-butoxy)carbonyl]amino}ethyl)-1H-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD18459235
  • Cas Number : 1211504-22-1
  • Molecular Weight : 256.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-94591

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-94591
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart