ENA514233970 Enamine

6-oxo-3-phenylpiperidine-3-carbonitrile

  • MDL number MFCD30180306
  • Cas Number : 1955541-39-5
  • Molecular Weight : 200.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-218673

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-218673
Availability details for
Quantity
  • Availability
  • Add to Cart