ENA514233973 Enamine

methyl 3-[(chlorosulfonyl)methyl]oxetane-3-carboxylate

  • MDL number MFCD30180317
  • Cas Number : 1955519-75-1
  • Molecular Weight : 228.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-244904

Safety Information

signalword Danger
pcodes P260,P264,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P363,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-244904
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart