ENA514233998 Enamine

tert-butyl 4-carbamothioyloxane-4-carboxylate

  • MDL number MFCD30180373
  • Cas Number : 1955520-94-1
  • Molecular Weight : 245.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-260086

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-260086
Availability details for
Quantity
  • Availability
  • Add to Cart