ENA514234128 Enamine

5-(4-bromophenyl)-3H,4H-thieno[2,3-d]pyrimidin-4-one

  • MDL number MFCD00435057
  • Cas Number : 307512-24-9
  • Molecular Weight : 307.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-17108

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-17108
Availability details for
Quantity
  • Availability
  • Add to Cart