ENA514234321 Enamine

1-fluoro-3-(2-methoxyethenyl)benzene

  • MDL number MFCD18835513
  • Cas Number : 438585-93-4
  • Molecular Weight : 152.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-214019

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214019
Availability details for
Quantity
  • Availability
  • Add to Cart