ENA514234339 Enamine

(3-chloro-5-fluorophenyl)hydrazine

  • MDL number MFCD11520842
  • Cas Number : 1260890-58-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-108024

Safety Information

signalword Warning
pcodes P201,P202,P261,P264,P270,P271,P280,P281,P301+P312,P302+P352,P304+P340,P305+P351+P338,P308+P313,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
hcodes Carc Cat 3,Xi,Xn
MSDS https://www.enaminestore.com/catalog/EN300-108024
pictograms GHS07,GHS08
hstatements H302,H315,H319,H335,H351
Availability details for
Quantity
  • Availability
  • Add to Cart