ENA514234350 Enamine

tert-butyl 4-aminopyridine-2-carboxylate

  • MDL number MFCD13196668
  • Cas Number : 868171-68-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-219405

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-219405
Availability details for
Quantity
  • Availability
  • Add to Cart