ENA514234369 Enamine

2-(2,6-dimethylphenyl)ethan-1-ol

  • MDL number MFCD09926174
  • Cas Number : 30595-80-3
  • Molecular Weight : 150.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-225837

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-225837
Availability details for
Quantity
  • Availability
  • Add to Cart