ENA514234382 Enamine

3-(bromomethyl)-3-(propan-2-yloxy)oxolane

  • MDL number MFCD30180316
  • Cas Number : 1935287-78-7
  • Molecular Weight : 223.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-243413

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-243413
Availability details for
Quantity
  • Availability
  • Add to Cart