ENA514234398 Enamine

decahydro-1,6-naphthyridin-2-one hydrochloride

  • MDL number MFCD30180256
  • Cas Number : 1955531-43-7
  • Molecular Weight : 190.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-109828

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109828
Availability details for
Quantity
  • Availability
  • Add to Cart