ENA514234406 Enamine

{5H,6H,7H,8H,9H-[1,2,3,4]tetrazolo[1,5-a]azepin-7-yl}methanol

  • MDL number MFCD30180325
  • Cas Number : 1936339-83-1
  • Molecular Weight : 168.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-248651

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-248651
Availability details for
Quantity
  • Availability
  • Add to Cart